Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM2 Antibody (MCM2/3678), FITC, Novus Biologicals™
SDP

Catalog No. NB155166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155166 Supplier Novus Biologicals Supplier No. NBP314118F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MCM2 Monoclonal antibody specifically detects MCM2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MCM2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MCM2/3678
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin)
Host Species Mouse
Immunogen Recombinant fragment (aa 650-750) of human MCM2 protein (exact sequence is proprietary) (Uniprot: P49736)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4171
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.