Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM3 Antibody, Novus Biologicals™
SDP

Catalog No. NB100289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100289 Supplier Novus Biologicals Supplier No. NB100289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

MCM3 Polyclonal specifically detects MCM3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen MCM3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:1000 to 1:5000, Immunoprecipitation 2-5 ug//mg lysate, Immunohistochemistry-Paraffin 1:1000 to 1:5000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias cervical cancer proto-oncogene 5, DNA polymerase alpha holoenzyme-associated protein P1, DNA replication factor MCM3, DNA replication licensing factor MCM3, EC 3.6.4.12, HCC5, hRlf beta subunit, MCM3 minichromosome maintenance deficient 3, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae), MGC1157, minichromosome maintenance complex component 3, minichromosome maintenance deficient (S. cerevisiae) 3, minichromosome maintenance deficient 3, P1.h, p102, P1-MCM3, replication licensing factor, beta subunit, RLF subunit beta, RLFB
Gene Symbols MCM3
Host Species Rabbit
Immunogen A 38-amino acid synthetic peptide, which represented a portion of human Minichromosome Maintenance Deficient 3 encoded in part by exons 10 and 11 (LocusLink ID 4172).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4172
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.