Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MCM4 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MCM4 Polyclonal specifically detects MCM4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
MCM4 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers | |
CDC21, CDC21 homolog, CDC54, DNA replication licensing factor MCM4, EC 3.6.4.12, hCdc21, homolog of S. pombe cell devision cycle 21, MCM4 minichromosome maintenance deficient 4, MCM4 minichromosome maintenance deficient 4 (S. cerevisiae), MGC33310, minichromosome maintenance complex component 4, minichromosome maintenance deficient 4, P1-Cdc21 | |
MCM4 | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
RUO | |
Human | |
4173 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SSPPQMHSSAIPLDFDVSSPLTYGTPSSRVEGTPRSGVRGTPVRQRPDLGSAQKGLQVDLQSDGAAAEDIVASEQSLGQKLVIWGTDVNVAACKENFQRFLQRFIDPLAKEEENVG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title