Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM6 Antibody (MCM6/2999), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB011307 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011307 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011307 Supplier Novus Biologicals Supplier No. NBP279862FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCM6 Monoclonal specifically detects MCM6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MCM6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MCM6/2999
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM
Gene Symbols MCM6
Host Species Mouse
Immunogen Recombinant fragment (around aa 228-368) of human MCM6 protein (exact sequence is proprietary) (Uniprot: Q14566)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.