Learn More
Description
Specifications
Specifications
| Antigen | MCM6 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | MCM6/3000 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | Q14566 |
| Gene Alias | DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
