Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Antibody (2068C), DyLight 550, R&D Systems™
SDP

Catalog No. MAB92171R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB92171R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB92171R Supplier Novus Biologicals Supplier No. MAB92171R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen MCM7
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2068C
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Gene Symbols MCM7
Host Species Rabbit
Immunogen Partial recombinant human MCM-7 protein (amino acids 188-328) [UniProt P33993]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4176
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.