Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Antibody (MCM7/1468), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP29611A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP29611A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP29611A30 Supplier Novus Biologicals Supplier No. NBP259611AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MCM7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MCM7/1468
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Gene Symbols MCM7
Host Species Mouse
Immunogen Recombinant human MCM7 protein fragment (aa195-319) (Uniprot: P33993)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4176
Test Specificity The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. MCM7 is one of the highly conserved mini-chromosome maintenance proteins (MCM) that is essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumor suppressor protein RB1/RB.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.