Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Antibody (MCM7/2832R), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB380769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380769 Supplier Novus Biologicals Supplier No. NBP308405AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen MCM7
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MCM7/2832R
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Host Species Rabbit
Immunogen Recombinant human MCM7 protein fragment (aa195-319) (Uniprot: P33993)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4176
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.