Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Mouse anti-Human, Alexa Fluor 647, Clone: 118H7, Novus Biologicals™
SDP

Catalog No. NB163045 Shop All R&D Systems Products
missing translation for 'changeView'
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. NB163045 Supplier Novus Biologicals missing translation for 'supplierNo' NBP260247AF647
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Mouse Monoclonal Antibody

MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen MCM7
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 118H7
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Host Species Mouse
Immunogen Recombinant human MCM7 (1-414aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4176
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.