Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCMBP Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304571100UL
Description
MCMBP Polyclonal specifically detects MCMBP in Human, Mouse samples. It is validated for Western Blot.Specifications
MCMBP | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
79892 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
C10orf119, FLJ13081, MCM-binding protein, MCM-BP, minichromosome maintenance complex binding protein, mini-chromosome maintenance complex-binding protein, minichromosome maintenance complex-binding protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human MCMBP (NP_079110.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction