Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCMBP Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MCMBP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MCMBP Polyclonal specifically detects MCMBP in Human, Mouse samples. It is validated for Western Blot.Specifications
MCMBP | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
79892 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
C10orf119, FLJ13081, MCM-binding protein, MCM-BP, minichromosome maintenance complex binding protein, mini-chromosome maintenance complex-binding protein, minichromosome maintenance complex-binding protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human MCMBP (NP_079110.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title