Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ McN-A 343
GREENER_CHOICESDP

Catalog No. 138410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-841-0 10 mg
13-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-841-0 Supplier Tocris Bioscience™ Supplier No. 1384/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective M1 muscarinic agonist

Selective muscarinic M1 receptor agonist. Selectivity for M1 over other muscarinic receptor types appears to arise from a high efficacy at M1 receptors.

Chemical Identifiers

CAS 55-45-8
Molecular Formula C14H18Cl2N2O2

Specifications

Quantity 10 mg
Formula Weight 317.21
Purity >99%
Molecular Weight (g/mol) 317.21
Chemical Name or Material McN-A 343
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.