Learn More
MCP-5 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$2175.00
Specifications
Antigen | MCP-5 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-271-8467
![]() |
Invitrogen™
500P61BT500UG |
500 μg |
Each of 1 for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-5 (CCL12). Anti-Murine MCP-5 (CCL12)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mMCP-5 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine MCP-5 (500-P61) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mMCP-5. Western Blot: To detect mMCP-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mMCP-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P61BT-1MG will be provided as 2 x 500 μg
Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.Specifications
MCP-5 | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Mouse | |
Q62401 | |
20293 | |
E.coli-derived Recombinant Murine MCP-5 (CCL12). | |
Antigen affinity chromatography | |
Ccl12 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
C-C motif chemokine 12; Ccl12; chemokine (C-C motif) ligand 12; MCP-1-related chemokine; Mcp5; MCP-5; Monocyte chemoattractant protein 5; monocyte chemotactic protein 5; Scya12; small inducible cytokine A12; small-inducible cytokine A12 | |
Ccl12 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.