Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT1/SLC16A1 Antibody (882616), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB373291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373291 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373291 Supplier Novus Biologicals Supplier No. FAB8275K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCT1/SLC16A1 Monoclonal specifically detects MCT1/SLC16A1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MCT1/SLC16A1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 882616
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MCT1/SLC16A1, Met1-Val500, Accession # P53985
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6566
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.