Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT1/SLC16A1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP159656SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159656SS 0.025 mg
NBP159656 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP159656SS Supplier Novus Biologicals Supplier No. NBP159656SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

MCT1/SLC16A1 Polyclonal specifically detects MCT1/SLC16A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen MCT1/SLC16A1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.5 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunohistochemistry-Paraffin 1:200, Knockdown Validated
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P53985
Gene Alias FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Symbols SLC16A1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Monocarboxylic acid transporter 1 protein (between residues 200-300) [UniProt P53985]
Molecular Weight of Antigen 54 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6566
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.