Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093883 20 μL
NB093882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093883 Supplier Novus Biologicals Supplier No. NBP30298720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCT2 Polyclonal specifically detects MCT2 in Human samples. It is validated for Western Blot.

Specifications

Antigen MCT2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias MCT 2, MCT2solute carrier family 16 (monocarboxylic acid transporters), member 7, monocarboxylate transporter 2, Solute carrier family 16 member 7, solute carrier family 16, member 7 (monocarboxylic acid transporter 2)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human MCT2 (NP_001257551.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9194
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.