Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT3/SLC16A8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151792 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151792 25 μg
NB151791 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151792 Supplier Novus Biologicals Supplier No. NBP31191925UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCT3/SLC16A8 Polyclonal antibody specifically detects MCT3/SLC16A8 in Human samples. It is validated for Western Blot

Specifications

Antigen MCT3/SLC16A8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400
Formulation PBS
Gene Alias MCT3MCT 3, monocarboxylate transporter 3, REMP, solute carrier 16 (monocarboxylic acid transporters), member 8, Solute carrier family 16 member 8, solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Host Species Rabbit
Immunogen Synthetic peptide derived from MCT3/SLC16A8 protein. (Uniprot: O95907)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23539
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.