Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MD-1/LY86 Antibody (241), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB072874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072874 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072874 Supplier Novus Biologicals Supplier No. NBP290386AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MD-1/LY86 Monoclonal specifically detects MD-1/LY86 in Mouse samples. It is validated for ELISA.

Specifications

Antigen MD-1/LY86
Applications ELISA
Classification Monoclonal
Clone 241
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MD-1/LY86 (Met 1-Ser 162; NP_034875.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9450
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.