Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MD-2 Mouse anti-Human, Mouse, Rat, HRP, Clone: 1A2E3, Novus Biologicals™
SDP

Catalog No. NB206494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206494 Supplier Novus Biologicals Supplier No. NBP175513H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MD-2 Monoclonal antibody specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MD-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A2E3
Conjugate HRP
Formulation PBS
Gene Alias ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2
Host Species Mouse
Immunogen Antibody was raised against a recombinant protein corresponding to amino acids 2 to 160 of human MD-2.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23643
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.