Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MDA-MB-361 Whole Cell Lysate
SDP

Catalog No. NBP20396201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP20396201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP20396201 Supplier Novus Biologicals™ Supplier No. NBP2039620.1MG

MDA-MB-361 (Human Breast Adenocarcinoma) whole breast cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: DMEM/Hams F12 and 10% NCS (newborn Calf serum). Lysates are guaranteed for 6 months from date of receipt.

The MDA-MB-361 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Tissue MDA-MB-361
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity MDA-MB-361 (Human Breast Adenocarcinoma)
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.