Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDA5 Antibody (1B6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197526
Description
MDA5 Monoclonal specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| MDA5 | |
| Monoclonal | |
| Unconjugated | |
| CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
| Mouse | |
| 50 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 64135 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 1B6 | |
| Western Blot 1:1000, ELISA | |
| IFIH1 | |
| Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction