Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDA5 Antibody (1B6), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | MDA5 |
---|---|
Clone | 1B6 |
Dilution | Western Blot 1:1000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
MDA5 Monoclonal specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
MDA5 | |
Western Blot 1:1000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
64135 | |
Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1B6 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
IFIH1 | |
IgG1 | |
Protein G purified | |
50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title