Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDA5 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB195748 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB195748 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB195748 Supplier Novus Biologicals Supplier No. NBP176761AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDA5 Polyclonal antibody specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MDA5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human MDA5. The immunogen is located within amino acids 330 - 380 of MDA5. Amino Acid Squence: AKDHLDKKKKASEPGK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 64135
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.