Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDA5 Antibody (OTI8C10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272604A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272604A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272604A32 Supplier Novus Biologicals Supplier No. NBP272604AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDA5 Monoclonal specifically detects MDA5 in Human samples. It is validated for Western Blot.

Specifications

Antigen MDA5
Applications Western Blot
Classification Monoclonal
Clone OTI8C10
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116
Gene Symbols IFIH1
Host Species Mouse
Immunogen Full length recombinant protein of human MDA5 (NP_071451) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64135
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.