Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB100395AFX Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100395AFX 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100395AFX Supplier Novus Biologicals Supplier No. NB100395AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDC1 Polyclonal specifically detects MDC1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen MDC1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
Gene Symbols MDC1
Host Species Rabbit
Immunogen A synthetic peptide, which represented a portion of human Mediator of DNA Damage Checkpoint Protein 1 encoded within exon 5 (NP_055456.1).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9656
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.