Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDC1 Antibody (P2B11), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212890UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212890UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212890UV Supplier Novus Biologicals Supplier No. NBP212890UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDC1 Monoclonal specifically detects MDC1 in Human, Mouse, Bovine, Non-human Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MDC1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone P2B11
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
Gene Symbols MDC1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9656
Test Specificity Detects approx 184kDa. This antibody recognizes MDC1 at and around the N-terminus.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.