Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDGA2 Antibody (803732), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB368384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368384 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368384 Supplier Novus Biologicals Supplier No. FAB5184W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDGA2 Monoclonal specifically detects MDGA2 in Human, Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MDGA2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 803732
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MAM domain containing glycosylphosphatidylinositol anchor 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MDGA2, Gln21-Asp934, Accession # Q7Z553
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161357
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.