Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP17771401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17771401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17771401 Supplier Novus Biologicals Supplier No. NBP1777140.1MG

Sheep Polyclonal Antibody

MDH1 Polyclonal specifically detects MDH1 in Porcine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen MDH1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias cytoplasmic, Cytosolic malate dehydrogenase, malate dehydrogenase 1, NAD (soluble), MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase
Gene Symbols MDH1
Host Species Sheep
Immunogen Malate Dehydrogenase [Pig Heart]
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 4190
Target Species Pig, Sheep
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.