Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH1 Antibody (CPTC-MDH1-1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB387060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387060 Supplier Novus Biologicals Supplier No. NBP308427AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDH1 Monoclonal antibody specifically detects MDH1 in Human samples. It is validated for Western Blot

Specifications

Antigen MDH1
Applications Western Blot
Classification Monoclonal
Clone CPTC-MDH1-1
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase
Host Species Mouse
Immunogen Recombinant human full-length MDH1 protein (Uniprot: P40925)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 4190
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.