Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Mdivi 1
GREENER_CHOICE

Catalog No. p-4816960
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective dynamin inhibitor; attenuates mitochondrial division and apoptosis

Mdivi 1 is a selective inhibitor of dynamin GTPase (IC50 = 1 - 10 μM); attenuates Dnm1- and Drp1-mediated mitochondrial division. Mdivi 1 inhibits mitochondrial outer membrane permeabilization (MOMP) and attenuates apoptosis. In cancer cell lines, Mdivi 1 reduces cell proliferation; in an animal model of ischemic retina Mdivi 1 treatment blocks apoptosis. In a cell model, Mdivi 1 protects against mutant huntington-induced damage and mitochondrial dysfunction. Mdivi 1 also inhibits autophagy and protects neurons from excitotoxicity.

Chemical Identifiers

CAS 338967-87-6
Molecular Formula C15H10Cl2N2O2S

Specifications

Quantity 10 mg
Formula Weight 353.22
Purity >98%
Molecular Weight (g/mol) 353.217
Chemical Name or Material Dynamin Inhibitors
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.