Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDL-1/CLEC5A Antibody (226402), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314315 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314315 Supplier Novus Biologicals Supplier No. FAB1639PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MDL-1/CLEC5A Monoclonal specifically detects MDL-1/CLEC5A in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MDL-1/CLEC5A
Applications Flow Cytometry
Classification Monoclonal
Clone 226402
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse MDL-1/CLEC5A
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 23601
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.