Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDL-1/CLEC5A Antibody (283834), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB365385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365385 Supplier Novus Biologicals Supplier No. FAB2384W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDL-1/CLEC5A Monoclonal specifically detects MDL-1/CLEC5A in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen MDL-1/CLEC5A
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 283834
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MDL-1/CLEC5A, Tyr26-Lys188, Accession # Q9NY25
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 23601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.