Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MDL 100907
GREENER_CHOICE

Catalog No. p-4835945
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT2A antagonist

MDL 100907 is a potent and selective 5-HT2A receptor antagonist (Ki = 0.36 nM, IC50 = 3.3-5.1 nM). MDL 100907 exhibits > 80-fold selectivity for 5-HT2A receptors over other serotonergic receptor subtypes. MDL 100907 is neuroprotective; protects cultured dopaminergic neurons from MPP+ induced toxicity. Antipsychotic agent in vivo.

Chemical Identifiers

CAS 139290-65-6
Molecular Formula C22H28FNO3

Specifications

Quantity 10 mg
Formula Weight 373.46
Molecular Weight (g/mol) 373.47
Chemical Name or Material MDL 100907
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.