Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MDL 11,939
GREENER_CHOICE

Catalog No. p-4783624
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT2A antagonist

Orally active 5-HT2A receptor antagonist; displays selectivity for 5-HT2A receptors over 5-HT2C receptors (Ki values are 0.54, 2.5, 81.6 and ∽10,000 nM at rabbit 5-HT2A, human 5-HT2A, rabbit 5-HT2C and human 5-HT2C receptors respectively).

Chemical Identifiers

CAS 107703-78-6
Molecular Formula C20H25NO

Specifications

Quantity 10 mg
Formula Weight 295.42
Purity >99%
Molecular Weight (g/mol) 295.426
Chemical Name or Material MDL 11, 939
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.