Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MDL 72222
GREENER_CHOICESDP

Catalog No. 064050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-405-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-405-0 Supplier Tocris Bioscience™ Supplier No. 0640/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

5-HT3 antagonist

5-HT3 antagonist.

Chemical Identifiers

CAS 40796-97-2
Molecular Formula C15H17Cl2NO2

Specifications

Quantity 50 mg
Formula Weight 314.21
Purity >99%
Molecular Weight (g/mol) 314.206
Chemical Name or Material MDL 72222
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.