Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | MDM1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MDM1 Polyclonal specifically detects MDM1 in Human samples. It is validated for Western Blot.Specifications
| MDM1 | |
| Polyclonal | |
| Rabbit | |
| B2RB22 | |
| 56890 | |
| Synthetic peptides corresponding to MDM1(Mitochondrial deafness modifier 1) The peptide sequence was selected from the N terminal of MDM1. Peptide sequence GLRSDQLGITKEPSFISKRRVPYHDPQISKSLEWNGAISESNVVASPEPE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ95264, Mdm1 nuclear protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 1, p53 binding protein, Mdm4, transformed 3T3 cell double minute 1, p53 binding protein (mouse), nuclear protein double minute 1, nuclear protein MDM1 | |
| MDM1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title