Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2/HDM2 Antibody (HDM2-323), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB6001312R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001312R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001312R Supplier Novus Biologicals Supplier No. NB6001312R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MDM2/HDM2
Applications Western Blot, ELISA
Classification Monoclonal
Clone HDM2-323
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Gene Symbols MDM2
Host Species Mouse
Immunogen Synthetic peptide conjugated to KLH, corresponding to amino acids 389-402 of Human MDM2.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4193
Test Specificity MDM2 (HDM2-323)
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.