Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2/HDM2, p Ser185 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB151192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151192 Supplier Novus Biologicals Supplier No. NB1001683DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDM2/HDM2 Polyclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for ELISA, Immunoprecipitation, Western Blot.

Specifications

Antigen MDM2/HDM2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Host Species Rabbit
Immunogen This affinity purified MDM2/HDM2 [p Ser185] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 175-200 of mouse MDM2/HDM2/HDM2. (Uniprot: P23804)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4193
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.