Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDMX Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB041749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041749 Supplier Novus Biologicals Supplier No. NBP297044DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDMX Polyclonal specifically detects MDMX in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MDMX
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant MDMX (NP_002384.2; Met1-Asp134).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4194
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.