Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1/ABCB1 Antibody (OTI3B2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP272606B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272606B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272606B Supplier Novus Biologicals Supplier No. NBP272606B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen MDR1/ABCB1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3B2
Conjugate Biotin
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Gene Symbols ABCB1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 347-710 of human MDR1/ABCB1 (NP_000918) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 5243.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.