Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ME2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB325826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325826 Supplier Novus Biologicals Supplier No. NBP299375MFV500

Rabbit Polyclonal Antibody

ME2 Polyclonal antibody specifically detects ME2 in Human samples. It is validated for Western Blot

Specifications

Antigen ME2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1, EC 1.1.1.38, malate dehydrogenase, Malic enzyme 2, malic enzyme 2, NAD(+)-dependent, mitochondrial, NAD-dependent malic enzyme, mitochondrial, NAD-ME, ODS1, pyruvic-malic carboxylase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ME2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 4200
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.