Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEA1 Mouse anti-Human, Rat, Clone: 1H8C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MEA1 |
---|---|
Clone | 1H8C2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6895
![]() ![]() |
Proteintech Group Inc
664721IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MEA1 gene ontology annotations related to this gene include spermatogenesis.Specifications
MEA1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HYS, male enhanced antigen 1, MEA, MEA 1, MEA1 | |
MEA1 | |
IgG1 | |
Protein G | |
Antibody |
1H8C2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q16626, Q5FVH7 | |
4201, 685131 | |
MEA1 Fusion Protein Ag0375 | |
Primary | |
-20°C | |
MEA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title