Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Measles nucleoprotein Antibody (83KKII) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1103507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1103507 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1103507 Supplier Novus Biologicals Supplier No. NB1103507
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Measles nucleoprotein Monoclonal specifically detects Measles nucleoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Measles nucleoprotein
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 83KKII
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias Measles nucleocapsid protein, N, NP, NP protein, Nucleocapsid protein, Nucleoprotein, Protein N
Host Species Mouse
Immunogen Recombinant measles NP against Edmonston strain measles virus and is known to react with all known strains of measles.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity NB110-3507 recognizes the measles nucleoprotein by capture EIA.
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.