Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MeCP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056326 0.1 mg
NB10056326T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056326 Supplier Novus Biologicals Supplier No. NB10056326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

MeCP2 Polyclonal specifically detects MeCP2 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MeCP2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. P51608
Gene Alias AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16
Gene Symbols MECP2
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 11-25 and 181-195 of human MeCP2.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4204
Test Specificity In HeLa, a 53 kDa band is observed.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.