Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MeCP2, p Ser80 Antibody, Novus Biologicals™
SDP

Catalog No. NB387847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387847 Supplier Novus Biologicals Supplier No. NBP323406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MeCP2 Polyclonal antibody specifically detects MeCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MeCP2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/ Immunofluorescence 1:250
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16
Host Species Rabbit
Immunogen phospho-MeCP2 (Ser-80) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 80 in mouse MeCP2. This peptide sequence is highly conserved in rat and human MeCP2, and has low homology to other nuclear proteins. [UniProt# Q9Z2D6]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4204
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.