Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED12 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB1002357IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002357IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002357IR Supplier Novus Biologicals Supplier No. NB1002357IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED12 Polyclonal specifically detects MED12 in Human, Mouse, Primate samples. It is validated for ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Western Blot (Negative).

Specifications

Antigen MED12
Applications ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Western Blot (Negative)
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARC240, CAG repeat protein 45, CAGH45trinucleotide repeat containing 11 (THR-associated protein, 230kDa subunit), HOPAFGS1, KIAA0192FG syndrome 1, mediator complex subunit 12OPA-containing protein, OKSsubunit 12 homolog, thyroid hormone receptor-associated protein, 230 kDa subunit, Trap230, TRAP230mediator of RNA polymerase II transcription, subunit 12 homolog (S. cerevisiae), trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit), Trinucleotide repeat-containing gene 11 protein
Gene Symbols MED12
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2150 and the C-terminus (residue 2212) of human Mediator of RNA Polymerase II Transcription, subunit 12 (NP_005111.1).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 9968
Target Species Human, Mouse, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.