Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED13 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060641 Supplier Novus Biologicals Supplier No. NB10060641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED13 Polyclonal specifically detects MED13 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen MED13
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_005112.2
Gene Alias Activator-recruited cofactor 250 kDa component, KIAA0593HSPC221, mediator complex subunit 13ARC250, mediator of RNA polymerase II transcription, subunit 13 homolog, THRAP1mediator of RNA polymerase II transcription subunit 13, thyroid hormone receptor associated protein 1, Thyroid hormone receptor-associated protein 1, Thyroid hormone receptor-associated protein complex 240 kDa component, thyroid hormone receptor-associated protein complex component TRAP240, thyroid hormone receptor-associated protein, 240 kDa subunit, Trap240, TRAP240DRIP250, Vitamin D3 receptor-interacting protein complex component DRIP250
Gene Symbols MED13
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human thyroid hormone receptor associated protein 1 (thyroid hormone receptor-associated protein, 240 kDa) using the numbering given in entry NP_005112.2 (GeneID 996
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9969
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.