Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP256318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP256318 100 μL
NB403735 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP256318 Supplier Novus Biologicals Supplier No. NBP256318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED14 Polyclonal specifically detects MED14 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MED14
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias Activator-recruited cofactor 150 kDa component, ARC150, Cofactor required for Sp1 transcriptional activation subunit 2, cofactor required for Sp1 transcriptional activation, subunit 2 (150kD), CRSP150, CRSP2cofactor required for Sp1 transcriptional activation, subunit 2, 150kDa, CSRP, CXorf4MGC104513, EXLM1CRSP complex subunit 2, human homolog of yeast RGR1, mediator complex subunit 14DRIP150, mediator of RNA polymerase II transcription subunit 14, RGR1RGR1 homolog, Thyroid hormone receptor-associated protein complex 170 kDa component, thyroid hormone receptor-associated protein complex component TRAP170, transcriptional co-activator CRSP150, Transcriptional coactivator CRSP150, Trap170, TRAP170hRGR1, vitamin D receptor-interacting protein complex component DRIP150, Vitamin D3 receptor-interacting protein complex 150 kDa component
Gene Symbols MED14
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PIAPPGTPAVVLKSKMLFFLQLTQKTSVPPQEPVSIIVPIIYDMASGTTQQADIPRQQNSSVAAPMMVSNILKRFAEMNP
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9282
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.