Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094587 20 μL
NB094586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094587 Supplier Novus Biologicals Supplier No. NBP30505520UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED14 Polyclonal specifically detects MED14 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MED14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Activator-recruited cofactor 150 kDa component, ARC150, Cofactor required for Sp1 transcriptional activation subunit 2, cofactor required for Sp1 transcriptional activation, subunit 2 (150kD), CRSP150, CRSP2cofactor required for Sp1 transcriptional activation, subunit 2, 150kDa, CSRP, CXorf4MGC104513, EXLM1CRSP complex subunit 2, human homolog of yeast RGR1, mediator complex subunit 14DRIP150, mediator of RNA polymerase II transcription subunit 14, RGR1RGR1 homolog, Thyroid hormone receptor-associated protein complex 170 kDa component, thyroid hormone receptor-associated protein complex component TRAP170, transcriptional co-activator CRSP150, Transcriptional coactivator CRSP150, Trap170, TRAP170hRGR1, vitamin D receptor-interacting protein complex component DRIP150, Vitamin D3 receptor-interacting protein complex 150 kDa component
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 470-650 of human MED14 (NP_004220.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9282
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.