Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED15 Antibody (OTI1C11), PE, Novus Biologicals™
SDP

Catalog No. NB287939 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287939 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287939 Supplier Novus Biologicals Supplier No. NBP272618PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED15 Monoclonal antibody specifically detects MED15 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen MED15
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C11
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias ARC105Arc105, CAG7A, CTG repeat protein 7a, CTG7A, FLJ42282, FLJ42935, mediator complex subunit 15PC2 glutamine/Q-rich-associated protein, mediator of RNA polymerase II transcription subunit 15, PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associatedprotein, PC2-glutamine-rich-associated protein, PCQAPDKFZp686A2214, Positive cofactor 2 glutamine/Q-rich-associated protein, positive cofactor 2, glutamine/Q-rich-associated protein, TIG1DKFZp762B1216, TIG-1TPA-inducible gene 1 protein, TNRC7, TPA inducible gene-1, TPA inducible protein, trinucleotide repeat containing 7,105-kD, Trinucleotide repeat-containing gene 7 protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 393-679 of human MED15(NP_056973) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51586
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.